Learn More
Medchemexpress LLC GM-CSF Human HEK29 500ug | 500ug
SDP
Catalog No. 5000195029
Encompass_Preferred
The GM-CSF protein functions as a key cytokine that promotes the growth and differentiation of hematopoietic precursor cells of different lineages such as granulocytes macrophages eosinophils and erythrocytes In its monomeric form GM-CSF acts as a signaling molecule orchestrating complex receptor assemblies GM-CSF Protein Human (HEK293) is the recombinant human-derived GM-CSF protein expressed by HEK293 with tag free
Catalog No. 50-001-95029 Supplier Medchemexpress LLC Supplier No. HYP78868500UG
The GM-CSF protein functions as a key cytokine that promotes the growth and differentiation of hematopoietic precursor cells of different lineages such as granulocytes macrophages eosinophils and erythrocytes In its monomeric form GM-CSF acts as a signaling molecule orchestrating complex receptor assemblies GM-CSF Protein Human (HEK293) is the recombinant human-derived GM-CSF protein expressed by HEK293 with tag free