Learn More
Medchemexpress LLC GM-CSF Human HEK29 50ug | 50ug
SDP
Catalog No. 5000194972
Encompass_Preferred
The GM-CSF protein functions as a key cytokine that promotes the growth and differentiation of hematopoietic precursor cells of different lineages such as granulocytes macrophages eosinophils and erythrocytes In its monomeric form GM-CSF acts as a signaling molecule orchestrating complex receptor assemblies GM-CSF Protein Human (HEK293) is the recombinant human-derived GM-CSF protein expressed by HEK293 with tag free
Catalog No. 50-001-94972 Supplier Medchemexpress LLC Supplier No. HYP7886850UG
The GM-CSF protein functions as a key cytokine that promotes the growth and differentiation of hematopoietic precursor cells of different lineages such as granulocytes macrophages eosinophils and erythrocytes In its monomeric form GM-CSF acts as a signaling molecule orchestrating complex receptor assemblies GM-CSF Protein Human (HEK293) is the recombinant human-derived GM-CSF protein expressed by HEK293 with tag free