Learn More
Medchemexpress LLC HY-P7361 10ug Medchemexpress, GM-CSF, Mouse CAS: Purity:>98%
SDP
Catalog No. 501934382
Encompass_Preferred
Medchemexpress, HY-P7361 10ug GM-CSF, Mouse CAS: Recombinant Mouse Granulocyte Macrophage Colony Stimulating Factor is a member of colony-stimulating factors which regulates proliferation, differentiation, and survival of hematopoietic cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4382 Supplier Medchemexpress LLC Supplier No. HYP736110UG
Medchemexpress, HY-P7361 10ug GM-CSF, Mouse CAS: Recombinant Mouse Granulocyte Macrophage Colony Stimulating Factor is a member of colony-stimulating factors which regulates proliferation, differentiation, and survival of hematopoietic cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.