Learn More
Medchemexpress LLC GM-CSF Mouse 500ug
SDP
Catalog No. 5000180717
Encompass_Preferred
GM-CSF Protein is a hematopoietic growth factor and immunomodulator By binding to specific receptors on the surface of target cells GM-CSF Protein activates intracellular signaling pathways to regulate cell proliferation differentiation maturation and function GM-CSF Protein plays an important role in the hematopoietic process and the immune system GM-CSF Protein Mouse is a recombinant GM-CSF Protein expressed by E coli without a tag[1][2][3][4]
Catalog No. 50-001-80717 Supplier Medchemexpress LLC Supplier No. HYP7361500UG
GM-CSF Protein is a hematopoietic growth factor and immunomodulator By binding to specific receptors on the surface of target cells GM-CSF Protein activates intracellular signaling pathways to regulate cell proliferation differentiation maturation and function GM-CSF Protein plays an important role in the hematopoietic process and the immune system GM-CSF Protein Mouse is a recombinant GM-CSF Protein expressed by E coli without a tag[1][2][3][4]