Learn More
Medchemexpress LLC GM-CSF Mouse HEK29 10ug
SDP
Catalog No. 5000180247
Encompass_Preferred
The GM-CSF protein acts as a potent cytokine that coordinates the growth and differentiation of hematopoietic precursor cells of different lineages including granulocytes macrophages eosinophils and erythrocytes Structurally GM-CSF exists as a monomer and its signaling is mediated through a dodecamer complex GM-CSF Protein Mouse (HEK293 Fc) is the recombinant mouse-derived GM-CSF protein expressed by HEK293 with N-hFc labeled tag
Catalog No. 50-001-80247 Supplier Medchemexpress LLC Supplier No. HYP7308210UG
The GM-CSF protein acts as a potent cytokine that coordinates the growth and differentiation of hematopoietic precursor cells of different lineages including granulocytes macrophages eosinophils and erythrocytes Structurally GM-CSF exists as a monomer and its signaling is mediated through a dodecamer complex GM-CSF Protein Mouse (HEK293 Fc) is the recombinant mouse-derived GM-CSF protein expressed by HEK293 with N-hFc labeled tag