Learn More
Enzo Life Sciences GM-CSF (mouse), (recombinant) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2530100
Encompass_Preferred
Alternative name: Granulocyte-macrophage colony stimulating factor, CSF-2, Pluripoietin-alpha, MGI1GM. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 10 mM acetic acid. Source: Produced in E. coli. Contains 125 amino acids. Long Term Storage: -80°C.
Catalog No. 50-201-4069