Learn More
Enzo Life Sciences GM-CSF (mouse), (recombinant) (5µg)
SDP
Supplier:  Enzo Life Sciences ADI908064005
Encompass_Preferred
Alternative name: CSF2, Colony stimulating factor 2 (granulocyte-macrophage). Purity: ≥98% (RP-HPLC and SDS-PAGE). Formulation: Lyophilized with no additives. Source: Produced in E. coli. Long Term Storage: +4°C. Applications: in vitro Assay.
Catalog No. 50-200-7091