Learn More
Genscript Corporation Antibody Protein Proteins Proteins 10 UG 1/EA
Supplier:  Genscript Corporation Z0299210
Encompass
Granulocyte Macrophage-Colony Stimulating Factor (GM-CSF) was initially characterized as a growth factor that can support the in vitro colony formation of granulocyte-macrophage progenitors. Granulocyte Macrophage-Colony Stimulating Factor (GM-CSF) is produced by a number of different cell types including activated T cells B cells macrophages mast cells endothelial cells and fibroblasts in response to cytokine of immune and inflammatory stimuli. Besides granulocyte-macrophage progenitors Granulocyte Macrophage-Colony Stimulating Factor (GM-CSF) is a growth factor for erythroid megakaryocyte and eosinophil progenitors. On mature hematopoietic monocytes/macrophages and eosinophils. Additionally Granulocyte Macrophage-Colony Stimulating Factor (GM-CSF) can stimulate the proliferation of a number of tumor cell lines including osteogenic sarcoma carcinoma and adenocarcinoma cell lines.
Catalog No. 50-252-9909