Learn More
Medchemexpress LLC GM-CSF Rat CHO 2ug
SDP
Catalog No. 5000197043
Encompass_Preferred
GM-CSF Protein, Rat (CHO) is a recombinant rat granulocyte-macrophage colony-stimulating factor expressed in Chinese hamster ovary (CHO) cells. It is supplied as a purified, biologically active, glycosylated full-length mature protein suitable for in vitro cell culture, bioassays, and functional studies. The product is manufactured to low endotoxin specifications and is provided in small pack sizes for sensitive research applications.
  • Produced in CHO cells for mammalian-type glycosylation.
  • Full-length mature sequence (A18-K144) representing native protein.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Low endotoxin level (<0.2 EU/μg) suitable for sensitive assays.
  • Predicted mass 14.6 kDa; apparent 16-26 kDa due to glycosylation.
  • Stable frozen; aliquot and store at -20 °C or -80 °C for extended storage.
Catalog No. 50-001-97043 Supplier Medchemexpress LLC Supplier No. HYP70942UG
GM-CSF Protein, Rat (CHO) is a recombinant rat granulocyte-macrophage colony-stimulating factor expressed in Chinese hamster ovary (CHO) cells. It is supplied as a purified, biologically active, glycosylated full-length mature protein suitable for in vitro cell culture, bioassays, and functional studies. The product is manufactured to low endotoxin specifications and is provided in small pack sizes for sensitive research applications.
  • Produced in CHO cells for mammalian-type glycosylation.
  • Full-length mature sequence (A18-K144) representing native protein.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Low endotoxin level (<0.2 EU/μg) suitable for sensitive assays.
  • Predicted mass 14.6 kDa; apparent 16-26 kDa due to glycosylation.
  • Stable frozen; aliquot and store at -20 °C or -80 °C for extended storage.