Learn More
Medchemexpress LLC GM-CSF Rat HEK293 10ug
SDP
Catalog No. 5000181382
Encompass_Preferred
GM-CSF Protein a cytokine promotes the growth and differentiation of hematopoietic precursor cells including granulocytes macrophages eosinophils and erythrocytes It functions as a monomer interacting with the GM-CSF receptor complex forming a dodecamer structure with two alpha two beta and two ligand subunits in head-to-head hexamers (By similarity) GM-CSF Protein Rat (HEK293 His) is the recombinant rat-derived GM-CSF protein expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-81382 Supplier Medchemexpress LLC Supplier No. HYP7262910UG
GM-CSF Protein a cytokine promotes the growth and differentiation of hematopoietic precursor cells including granulocytes macrophages eosinophils and erythrocytes It functions as a monomer interacting with the GM-CSF receptor complex forming a dodecamer structure with two alpha two beta and two ligand subunits in head-to-head hexamers (By similarity) GM-CSF Protein Rat (HEK293 His) is the recombinant rat-derived GM-CSF protein expressed by HEK293 with C-6 His labeled tag