Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ GM-CSF Recombinant Human Protein

Catalog No. CTP2013
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CTP2013 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CTP2013 Supplier Gibco™ Supplier No. CTP2013
Add to Cart
Add to Cart
Request a Quote

Recombinant Human GM-CSF

  • Involved in many biological responses including growth and development of granulocyte and macrophage progenitor cells
  • For stimulation and initiation of differentiation of myeloblasts and monoblasts, and chemotaxis of eosinophils
  • ED50 range = 0.02 to 0.1 ng/mL, determined by dose dependent proliferation of human TF-1 cells
  • Optimal concentration for individual application should be determined by dose response assay

Order Info

Shipping Condition: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Cell Culture
Molecular Weight (g/mol) 14 kDa
Purification Method Gel-Purified
Quantity 1 mg
Source Human
Gene Alias GM-CSF
Product Type Recombinant Protein
Species Human
Recombinant Recombinant
Content And Storage Lyophilized protein can be stored at 2-8°C dessicated. Once reconstituted, store in aliquots at ≤20°C. Avoid multiple freeze and thaw cycles.
Classification Carrier-Free
Activity 0.055 ng/mL
Bioassay Proliferation of TF1 cells
Endotoxin Level <0.1 ng/μg
Reactivity Human
Shipping Condition Wet Ice
Expression System E. coli
Protein Family Cytokines & Receptors
Protein Form Full Length, Recombinant, Ligand
Protein Subtype GM-CSF, G-CSF, & M-CSF
Research Category Clinical Research, Oncology, Stem Cell Research, Differentiation, Signal Transduction, Drug Development
Product Line Gibco
Purity or Quality Grade 95%
Show More Show Less
It looks like the recombinant protein vial I received from you is empty. Is this normal?

Gibco recombinant proteins are frequently formulated without carrier proteins or additives (e.g., BSA, HSA, sucrose, etc.) and no Gibco PeproTech recombinant protein contains a carrier protein. As a result, during lyophilization, the protein product may be deposited on the vial as a thin, and sometimes invisible, film instead of a pellet. The size of the pellet, if any, is not directly related to the quantity of the recombinant protein in the vial. Our quality control procedures assure that each vial contains the correct amount of product.

To ensure complete recovery of protein product, before opening a vial of lyophilized recombinant protein, we recommend centrifuging it in a microcentrifuge for 20-30 seconds to drive any protein that may be lodged in the cap or on the side to the bottom of the vial. After reconstitution, you can confirm the presence of product protein by running a small amount on SDS-PAGE. In general, a protein band with expected size should be visible with as little as 10 ng of protein loaded on an acrylamide gel.

Why does my recombinant protein not show activity in my own experiment?

Assay time is critical. Each assay needs to beoptimized and performed at the peak response time. Different cells may respond differently to a growth factor or cytokine. We suggest repeating our QC assay using same indicator cells as suggested in the manual to see if you can obtain a similar response. In addition, serum may be masking the response. Serum starvation might be needed for certain types of assays.

How should I store the reconstituted recombinant proteins?

Protein solutions are generally not very stable when frozen at low concentration. Upon freeze and thaw, some proteins in the solution may stick to the wall of the container, which results in significant reduction of protein concentration if the starting concentration was low. Therefore, carrier proteins are used to reduce such loss. The most commonly used carrier proteins include bovine serum albumin (BSA), human serum albumin (HSA), or fetal bovine serum (FBS). These carrier proteins are generally used at 0.1% concentration. As a rule of thumb, if the concentration of the recombinant protein is less than 0.5 mg/mL, it is a good idea to add some carrier protein

How do I store my lyophilized recombinant protein?

Lyophilized proteins can typically be stored at 2 to 8 degrees C for several weeks, or stored dessicated at -20 degrees C for long-term storage.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.