Learn More
Medchemexpress LLC GM-CSF Rhesus macaq 10ug
SDP
Catalog No. 5000183496
Encompass_Preferred
Recombinant GM-CSF protein derived from rhesus macaque, provided lyophilized for research use. The protein stimulates granulocyte and monocyte lineage cells and is intended for in vitro applications including cell culture, immune cell stimulation, and assay development. Expressed in E. coli and characterized for purity and molecular mass.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Predicted molecular mass of approximately 14 kDa.
  • Lyophilized formulation for convenient storage and reconstitution.
  • Suitable for in vitro cell culture and immune stimulation assays.
  • Stable when aliquoted and stored at -20°C or -80°C for extended periods.
Catalog No. 50-001-83496 Supplier Medchemexpress LLC Supplier No. HYP718410UG
Recombinant GM-CSF protein derived from rhesus macaque, provided lyophilized for research use. The protein stimulates granulocyte and monocyte lineage cells and is intended for in vitro applications including cell culture, immune cell stimulation, and assay development. Expressed in E. coli and characterized for purity and molecular mass.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Predicted molecular mass of approximately 14 kDa.
  • Lyophilized formulation for convenient storage and reconstitution.
  • Suitable for in vitro cell culture and immune stimulation assays.
  • Stable when aliquoted and stored at -20°C or -80°C for extended periods.