Learn More
ACROBiosystems ACROBiosystems ActiveMax Human GM-CSF Protein, Tag Free
Supplier:  ACROBiosystems GMFH42141MG
Encompass_Preferred
Recombinant Protein;1MG;ActiveMax Human GM-CSF, Tag Free (GMF-H4214) is expressed from human 293 cells (HEK293). It contains AA Ala 18 - Glu 144 (Accession # NP_000749.2).;This protein carries no "tag". The protein has a calculated MW of 14.5 kDa. The protein migrates as 18-26 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;The bio-activity was determined by dose-dependent stimulation of the proliferation of TF-1 cells. The ED50 < 0.1 ng/mL, corresponding to a specific activity of > 1x107 Unit/mg.
Catalog No. 50-195-7563