Learn More
ACROBiosystems ACROBiosystems Human GM-CSF R alpha Protein, His Tag (SPR verified)
Supplier:  ACROBiosystems GRAH52H7100UG
Encompass_Preferred
Recombinant Protein;100UG;Human GM-CSF R alpha, His Tag (SPR verified) (GRA-H52H7) is expressed from human 293 cells (HEK293). It contains AA Glu 23 - Gly 320 (Accession # P15509-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.4 kDa. The protein migrates as 60-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Human GM-CSF R alpha, His Tag (SPR verified) (Cat. No. GRA-H52H7) captured on CM5 Chip via anti-His antibody, can bind Human GM-CSF, Tag Free (Cat. No. GMF-H4214) with an affinity constant of 16.1 nM as determined in SPR assay (Biacore T200) (QC tested).
Catalog No. 50-195-7078