Learn More
Carbosynth LLC. GM1-Ganglioside sodium, 37758-47-7, MFCD00466936, 25 mg
SDP
Supplier:  Carbosynth LLC. OG0391825MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Chemical Formula: C73H131N3O31xNa, MW: 1546.82 (free acid basis), Synonyms: Gal(b1-3)GalNAc(b1-4)[Neu5Ac(a2-3)]Gal(b1-4)Glc(b1-1)Cers; Monosialoganglioside GM1; prototype ganglioside; monosialotetrahexosylganglioside.
Catalog No. NC1254572