Learn More
Enzo Life Sciences Ganglioside GM1. sodium salt (bovine brain) (10mg). CAS: 37758-47-7
SDP
Supplier:  Enzo Life Sciences ALX302001M010
Encompass_Preferred
Specific receptor for cholera toxin. Alternative name: GM1 . Na (bovine brain), Monosialoganglioside GM1 . Na (bovine brain). Purity: ≥98% (TLC). Long Term Storage: -20°C.
Catalog No. 50-200-8644