Learn More
Aviva Systems Biology Corp Gm13178 Antibody - C-terminal region (ARP44656_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44656_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Gm13178. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Gm437, OTTMUSG00000010750ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-156