Learn More
Aviva Systems Biology Corp GM14543 ANTIBODY 50UG/50UL
SDP
Supplier:  Aviva Systems Biology Corp ARP37658_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC436198. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the n terminal region of human LOC436198. Alternate names: Rhox7-like, OTTMUSG00000017128ApplicationWB. Species reactivity: Mouse
Catalog No. 50-284-681