Learn More
Cusabio Technology LLC GM2 ganglioside activator
Catalog No. 5000051137
Encompass_Preferred
This kit detects GM2A in human serum plasma urine tissue homogenates and cell lysates with a broad detection range of 15 6 pg/mL to 1000 pg/mL offering high sensitivity down to 3 9 pg/mL Utilizing a sandwich ELISA technique it enables quantitative analysis of GM2A levels within 1-5 hours of incubation time The assay requires a sample volume of 50-100 L and measures absorbance at 450 nm for accurate detection Designed for flexibility it accommodates diverse biological matrices while maintaining robust performance making it suitable for research applications requiring precise measurement of GM2A concentrations in multiple sample types
Catalog No. 50-000-51137 Supplier Cusabio Technology LLC Supplier No. CSBEL009565HU
This kit detects GM2A in human serum plasma urine tissue homogenates and cell lysates with a broad detection range of 15 6 pg/mL to 1000 pg/mL offering high sensitivity down to 3 9 pg/mL Utilizing a sandwich ELISA technique it enables quantitative analysis of GM2A levels within 1-5 hours of incubation time The assay requires a sample volume of 50-100 L and measures absorbance at 450 nm for accurate detection Designed for flexibility it accommodates diverse biological matrices while maintaining robust performance making it suitable for research applications requiring precise measurement of GM2A concentrations in multiple sample types