Learn More
Aviva Systems Biology Corp GM2A Antibody - N-terminal region (ARP46008_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46008_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GM2A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GM2A. Alternate names: SAP-3, GM2-APApplicationWB. Species reactivity: Human
Catalog No. 50-014-59174