Learn More
Aviva Systems Biology Corp GM2A Antibody - N-terminal region (ARP46007_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46007_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GM2A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GM2A. Alternate names: SAP-3, GM2-APApplicationWB. Species reactivity: Guinea Pig, Human
Catalog No. 50-285-235