Learn More
Enzo Life Sciences Ganglioside GM3. sodium salt (bovine brain) (1mg). CAS: 54827-14-4
SDP
Supplier:  Enzo Life Sciences ALX302005M001
Encompass_Preferred
The major ganglioside of the extra-nervous system. Inhibits growth of epidermal cells. Alternative name: GM3 . Na (bovine brain), Monosialoganglioside GM3 . Na (bovine brain). Purity: ≥98% (TLC). Appearance: Lyophilized. Solubility: Soluble in chlorform/methanol (2:1). Long Term Storage: -20°C.
Catalog No. 50-200-8645