Learn More
Aviva Systems Biology Corp GM5581 ANTIBODY 50UG/50UL
SDP
Supplier:  Aviva Systems Biology Corp ARP57916_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Gm5581. It was validated on Western Blot. . Alternate names: EG434094ApplicationWB. Species reactivity: Cow, Dog, Human, Pig
Catalog No. 50-286-412