Learn More
Aviva Systems Biology Corp PDLIM1 Antibody - middle region (ARP37659_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37659_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC545743. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human LOC545743. Alternate names: EG545743ApplicationWB. Species reactivity: Mouse
Catalog No. 50-284-682