Learn More
Aviva Systems Biology Corp GM6185 ANTIBODY 50UG/50UL
SDP
Supplier:  Aviva Systems Biology Corp ARP35520_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Gm6185. It was validated on Western Blot. . Alternate names: EG620839ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-284-465