Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences GMA (Hydroxyethyl Methacrylate), Low Acid 450 ML
SDP

Catalog No. 5024317
Encompass_Preferred

CAS #868-77-9 C₆H₁₀O₃ F.W. 130.14
Boiling point: 67°C

Inhibited with approximately 330ppm MEHQ. A water soluble monomer. Low Acid

Catalog No. 50-243-17 Supplier Electron Microscopy Sciences Supplier No. 16800
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS #868-77-9 C₆H₁₀O₃ F.W. 130.14
Boiling point: 67°C

Inhibited with approximately 330ppm MEHQ. A water soluble monomer. Low Acid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.