Learn More
Medchemexpress LLC HY-W041994 10mg Medchemexpress, GMBS CAS:80307-12-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW04199410MG
Encompass_Preferred
Medchemexpress, HY-W041994 10mg GMBS CAS:80307-12-6 GMBS is a heterobifunctional cross-linker. GMBS is often used to prepare antibody-enzyme and hapten-carrier protein conjugates. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4661