Learn More
Labelmaster Inc Genetically Modified Organism (GMO) Label, PVC-Free Film, 500 roll
SDP
Catalog No. NC1071690
Encompass_Preferred
When there is a requirement to ship Genetically Modified Organisms (GMO) by air, use Labelmaster's GMO UN3245 Labels. Printed on PVC-free vinyl, these shipping labels measure 50 x 50mm and are sold in rolls of 500.
Catalog No. NC1071690 Supplier Labelmaster Inc Supplier No. GMOV50
When there is a requirement to ship Genetically Modified Organisms (GMO) by air, use Labelmaster's GMO UN3245 Labels. Printed on PVC-free vinyl, these shipping labels measure 50 x 50mm and are sold in rolls of 500.