Learn More
Biolegend GMP APC anti-human CD28 [CD28.2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Supplier:  Biolegend 260250
Encompass_Preferred
GMP APC anti-human CD28 [CD28.2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-237-8309