Learn More
AnaSpec GMP Beta-Amyloid (1-42), Human, High Purity ≥97%, Lyophilized Form
Supplier:  AnaSpec ASGMP202761
Encompass
GMP Beta-Amyloid (1-42), human (Ammonium Salt) is a highly pure, lyophilized peptide critical for Alzheimer's disease research. Designed under stringent quality controls, it offers exceptional lot-to-lot reproducibility and consistency. Its compliance with regulatory standards enhances trust, making it ideal for developing diagnostics and preclinical studies. This unique peptide allows researchers to explore the mechanisms behind amyloid plaques, vital for understanding Alzheimer's pathology.
Catalog No. NC2378973