Learn More
CAPTIVATE BIO LLC GMP hPL heparin-required
Catalog No. 5000876342
Encompass_Preferred
PLTMax Human Platelet Lysate GMP 500mL from Captivate Bio is a GMP-grade is a growth factor-rich supplement that is a superior alternative to FBS PLTMax is used in the generation of MSCs and T-cells including neurology nephrology gastrointestinal disease wound repair and cardiology US origin
Catalog No. 50-008-76342 Supplier CAPTIVATE BIO LLC Supplier No. CBPLTMAX5G
PLTMax Human Platelet Lysate GMP 500mL from Captivate Bio is a GMP-grade is a growth factor-rich supplement that is a superior alternative to FBS PLTMax is used in the generation of MSCs and T-cells including neurology nephrology gastrointestinal disease wound repair and cardiology US origin