Learn More
CAPTIVATE BIO LLC GMP hPL xeno-free
Supplier: CAPTIVATE BIO LLC CBPLTGOLD1G
PLTGold Human Platelet Lysate GMP 100mL from Captivate Bio is a GMP-grade xeno-free heparin-free supplement for use cell therapy research Includes expanded endotoxin testing enhances genetic stability in MSC cultures and is ideal for generating cells in large scale bioreactors US origin