Learn More
CAPTIVATE BIO LLC GMP hPL xeno-free
Catalog No. 5000876340
Encompass_Preferred
PLTGold Human Platelet Lysate GMP 500mL from Captivate Bio is a GMP-grade xeno-free heparin-free supplement for use cell therapy research Includes expanded endotoxin testing enhances genetic stability in MSC cultures and is ideal for generating cells in large scale bioreactors US origin
Catalog No. 50-008-76340 Supplier CAPTIVATE BIO LLC Supplier No. CBPLTGOLD5G
PLTGold Human Platelet Lysate GMP 500mL from Captivate Bio is a GMP-grade xeno-free heparin-free supplement for use cell therapy research Includes expanded endotoxin testing enhances genetic stability in MSC cultures and is ideal for generating cells in large scale bioreactors US origin