Learn More
ACROBiosystems GMP Human FGF basic Protein
Supplier:  ACROBiosystems GMPFGCH171MG
Encompass_Preferred
GMP Human FGF basic Protein (GMP-FGCH17) is expressed from E. coli cells. It contains AA Pro 143 - Ser 288 (Accession # P09038-4).
Catalog No. 50-253-4990