Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems GMP Human IL-18 Protein, 1 mg, 18.2 kDa, >95% purity, >3.00 x 10^6 IU/mg, Lyophilized, Animal-Free

Catalog No. NC3628118
Encompass_Preferred

ACRO BIOSYSTEMS GMP Human IL-18 Protein is a high-quality, recombinant human interleukin-18 produced under stringent Good Manufacturing Practice (GMP) conditions. This potent proinflammatory cytokine plays a critical role in inducing interferon-gamma (IFN-γ) production and is manufactured with animal-free materials to support cell and gene therapy applications. The protein offers exceptional purity, high specific activity, and batch-to-batch consistency.

  • Manufactured under GMP conditions
  • High purity >95%
  • Specific activity >3.00 x 10^6 IU/mg
  • Storage stability of 5 years at -20°C to -70°C (lyophilized)
  • Host cell protein contamination <0.5 ng/μg
  • Endotoxin level <10 EU/mg
  • Mycoplasma-negative
  • Quality controlled under ISO 9001:2015 and ISO 13485:2016
  • FDA DMF filed
  • Animal-free material

Catalog No. NC3628118 Supplier ACROBiosystems Supplier No. GMPL18H161MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

ACRO BIOSYSTEMS GMP Human IL-18 Protein is a high-quality, recombinant human interleukin-18 produced under stringent Good Manufacturing Practice (GMP) conditions. This potent proinflammatory cytokine plays a critical role in inducing interferon-gamma (IFN-γ) production and is manufactured with animal-free materials to support cell and gene therapy applications. The protein offers exceptional purity, high specific activity, and batch-to-batch consistency.

  • Manufactured under GMP conditions
  • High purity >95%
  • Specific activity >3.00 x 10^6 IU/mg
  • Storage stability of 5 years at -20°C to -70°C (lyophilized)
  • Host cell protein contamination <0.5 ng/μg
  • Endotoxin level <10 EU/mg
  • Mycoplasma-negative
  • Quality controlled under ISO 9001:2015 and ISO 13485:2016
  • FDA DMF filed
  • Animal-free material

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.