Learn More
ACROBiosystems GMP human interleukin-2 (IL-2) protein, lyophilized, 1x10^6 IU
Catalog No. 502719568
Encompass_Preferred
GMP-grade human interleukin-2 (IL-2) protein supplied as a lyophilized preparation at a potency of 1x10^6 IU. Optimized for cell culture and immune cell stimulation assays, it provides consistent activity when stored and handled according to recommendations.
  • High specific activity (≥ 1.20x10^7 IU/mg).
  • Lyophilized format for extended shelf life.
  • Stable at -20°C for up to 5 years in lyophilized state.
  • After reconstitution, stable at -70°C for 12 months under sterile conditions.
  • Expressed in E. coli and 0.22 μm filtered.
  • Suitable for CTLL-2 proliferation and human PBMC culture.
Catalog No. 50-271-9568 Supplier ACROBiosystems Supplier No. GMPL02H141X106IU
 Restricted Item
Add to Cart
Add to Cart
GMP-grade human interleukin-2 (IL-2) protein supplied as a lyophilized preparation at a potency of 1x10^6 IU. Optimized for cell culture and immune cell stimulation assays, it provides consistent activity when stored and handled according to recommendations.
  • High specific activity (≥ 1.20x10^7 IU/mg).
  • Lyophilized format for extended shelf life.
  • Stable at -20°C for up to 5 years in lyophilized state.
  • After reconstitution, stable at -70°C for 12 months under sterile conditions.
  • Expressed in E. coli and 0.22 μm filtered.
  • Suitable for CTLL-2 proliferation and human PBMC culture.