Interleukin-6 (IL-6) is a multifunctional cytokine that regulates immune responses, hematopoiesis, and inflammatory reactions. Expressed in human HEK293 cells, this GMP-grade protein is produced in ISO 5 cleanrooms under a cGMP-compliant quality system to support clinical and large-scale manufacturing applications. It is designed to support the rapid expansion and viability of CD34+ hematopoietic stem cells and aids in the transition from mesoderm to hematopoietic commitment during iPSC differentiation.
- Validated specific activity of 1.00×108 IU/mg using the TF-1 cell line.
- High purity >95% as determined by SDS-PAGE.
- Low endotoxin levels <10 EU/mg and residual host cell DNA <0.02 ng/μg.
- Animal origin-free production and comprehensive regulatory support including DMF.
- Stable for 5 years when stored lyophilized at -20°C to -70°C.