Learn More
ACROBiosystems GMP Human Laminin 521 Protein
Catalog No. 502719565
Encompass_Preferred
Laminin 521 were expressed in Descemet's membrane and corneal endothelium. This laminin isoform significantly enhanced the in vitro adhesion and proliferation, and differentiation of HCECs. Laminin 511-E8 consists of the C-terminal end of the alpha 5, b
Catalog No. 50-271-9565 Supplier ACROBiosystems Supplier No. GMPLA5H241MG
Laminin 521 were expressed in Descemet's membrane and corneal endothelium. This laminin isoform significantly enhanced the in vitro adhesion and proliferation, and differentiation of HCECs. Laminin 511-E8 consists of the C-terminal end of the alpha 5, b