Learn More
ACROBiosystems GMP Human SCF Protein
Catalog No. 502534964
Encompass_Preferred
GMP Human SCF Protein (GMP-SCFH25) is expressed from human 293 cells (HEK293). It contains AA Glu 26 - Ala 189 (Accession # P21583-1).
Catalog No. 50-253-4964 Supplier ACROBiosystems Supplier No. GMPSCFH2550UG
GMP Human SCF Protein (GMP-SCFH25) is expressed from human 293 cells (HEK293). It contains AA Glu 26 - Ala 189 (Accession # P21583-1).