Learn More
ACROBiosystems GMP Human TNF-alpha Protein
Catalog No. 502534961
Encompass_Preferred
GMP Human TNF-alpha Protein (GMP-TNAH23) is expressed from human 293 cells (HEK293). It contains AA Val 77 - Leu 233 (Accession # P01375).
Catalog No. 50-253-4961 Supplier ACROBiosystems Supplier No. GMPTNAH2350UG
GMP Human TNF-alpha Protein (GMP-TNAH23) is expressed from human 293 cells (HEK293). It contains AA Val 77 - Leu 233 (Accession # P01375).