Learn More
Biolegend GMP PerCP/Cyanine5.5 anti-huma
Catalog No. 502745591
Encompass_Preferred
GMP PerCP/Cyanine5.5 anti-human CD27 [M-T271]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-274-5591 Supplier Biolegend Supplier No. 260404
GMP PerCP/Cyanine5.5 anti-human CD27 [M-T271]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests