Learn More
Biolegend GMP PerCP/Cyanine5.5 anti-huma
Catalog No. 503108010
Encompass_Preferred
GMP PerCP/Cyanine5 5 anti-human CD5 [L17F12] Isotype Mouse IgG2a Reactivity Human Apps FC Size 100 tests
Catalog No. 50-310-8010 Supplier Biolegend Supplier No. 260500
GMP PerCP/Cyanine5 5 anti-human CD5 [L17F12] Isotype Mouse IgG2a Reactivity Human Apps FC Size 100 tests