Learn More
Biolegend GMP PerCP/Cyanine5.5 anti-huma
Supplier:  Biolegend 260346
Encompass_Preferred
GMP PerCP/Cyanine5.5 anti-human CD16 [3G8]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-253-7959