Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci GMP Reductase 1 Recombinant Protein
SDP

Catalog No. 16592997
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-592-997 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-592-997 Supplier ProSci Supplier No. 916660050
Only null left
Add to Cart
Add to Cart

Recombinant Protein

This recombinant protein can be used for biological assays. For research use only.

GMP Reductase 1 (GMPR) is a member of the IMPDH/GMPR family. GMPR exists as a homotetramer and catalyzes the irreversible NADPH-dependent deamination of GMP to IMP. It functions in the conversion of nucleobase, nucleoside and nucleotide derivatives of G to A nucleotides, and in maintaining the intracellular balance of A and G nucleotides. GMP reductase gene expression may be regulated by MITF. At least two different alleles are known.

Specifications

Accession Number P36959
Gene ID (Entrez) 2766
Molecular Weight (g/mol) 38.5 kDa
Quantity 0.05 mg
Source Human Cells
Common Name guanosine monophosphate reductase
Gene Symbol GMPR
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag C-6 His tag
Sequence Met1-Ser345
Content And Storage Store at -20°C, stable for 6 months after receipt. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
Purity or Quality Grade >95% as determined by reducing SDS-PAGE. Endotoxin level less than 0.1 ng/μg (1 IEU/μg) as determined by LAL test.
Synonym GMP Reductase 1, Guanosine 5'-Monophosphate Oxidoreductase 1, Guanosine Monophosphate Reductase 1, GMPR, GMPR1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.