Learn More
Aviva Systems Biology Corp GMPR2 Antibody - C-terminal region (ARP48368_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48368_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GMPR2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human GMPR2. Alternate names: MGC15084, MGC830ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59806