Learn More
Medchemexpress LLC Gmprga Tfa | 98.1% | 5 MG
SDP
Catalog No. 5000399220
Encompass_Preferred
GMPRGA TFA is the trifluoroacetic acid (TFA) salt form of GMPRGA. It is known to recognize spAimR in the bacterial cytosol, a process that induces lysogeny.
  • TFA salt form
  • Recognizes spAimR in bacterial cytosol
  • Induces lysogeny
  • Functions as an inhibitor
  • Part of bacterial lysis-lysogeny studies
  • Classified as an antimicrobial peptide
Catalog No. 50-003-99220 Supplier Medchemexpress LLC Supplier No. HYP6011A5MG
GMPRGA TFA is the trifluoroacetic acid (TFA) salt form of GMPRGA. It is known to recognize spAimR in the bacterial cytosol, a process that induces lysogeny.
  • TFA salt form
  • Recognizes spAimR in bacterial cytosol
  • Induces lysogeny
  • Functions as an inhibitor
  • Part of bacterial lysis-lysogeny studies
  • Classified as an antimicrobial peptide