Learn More
Medchemexpress LLC Gmprga Tfa | 98.1% | 587.69 | 1 MG
SDP
Catalog No. 5000397111
Encompass_Preferred
GMPRGA (TFA) is the trifluoroacetate salt form of GMPRGA, a compound that plays a role in bacterial processes. It is known for its ability to recognize spAimR within the bacterial cytosol, which subsequently induces lysogeny. This product is intended for research applications only and is not suitable for human use.
  • Recognizes spAimR in the bacterial cytosol.
  • Induces lysogeny in bacteria.
  • For research purposes only.
Catalog No. 50-003-97111 Supplier Medchemexpress LLC Supplier No. HYP6011A1MG
GMPRGA (TFA) is the trifluoroacetate salt form of GMPRGA, a compound that plays a role in bacterial processes. It is known for its ability to recognize spAimR within the bacterial cytosol, which subsequently induces lysogeny. This product is intended for research applications only and is not suitable for human use.
  • Recognizes spAimR in the bacterial cytosol.
  • Induces lysogeny in bacteria.
  • For research purposes only.