Learn More
Cayman Chemical GMQ (hydrochloride), Purity: ≥95%, CAS: 5361-15-9, 5 mg
SDP
Catalog No. 502988159
Encompass_Preferred
An ASIC3 activator (EC50 = 67 µM for evoking rapid inward currents in ASIC3-expressing CHO cells at neutral pH); an inhibitor of the GABAA receptor (IC50 = 0.39 µM in primary rat hippocampal neurons); increases paw licking time in Asic3+/+ and, to a lesser extent, Asic3-/- mice at 100 µM
Catalog No. 50-298-8159 Supplier Cayman Chemical Supplier No. 297605 MG
An ASIC3 activator (EC50 = 67 µM for evoking rapid inward currents in ASIC3-expressing CHO cells at neutral pH); an inhibitor of the GABAA receptor (IC50 = 0.39 µM in primary rat hippocampal neurons); increases paw licking time in Asic3+/+ and, to a lesser extent, Asic3-/- mice at 100 µM