Learn More
Diagnostic Biosystems GMS GMS
Catalog No. 502407494
Encompass_Preferred
The Modified Gomori Methenamine-Silver Nitrate Stain (GMS Stain Kit) is intended for use in the histologic visualization of fungi, basement membrane and some opportunistic organisms such as Pneumocystis carinii. Pneumocystis carinii is an opportunistic pathogen that causes severe pulmonary disease in humans, dogs, rats, mice and other vertebrate species with acquired, induced, or inherited immune deficiency syndromes. In addition, this procedure will demonstrate Actinomyces and related species, Nocardia asteroids, and certain encapsulated bacteria.
Catalog No. 50-240-7494 Supplier Diagnostic Biosystems Supplier No. KT015
The Modified Gomori Methenamine-Silver Nitrate Stain (GMS Stain Kit) is intended for use in the histologic visualization of fungi, basement membrane and some opportunistic organisms such as Pneumocystis carinii. Pneumocystis carinii is an opportunistic pathogen that causes severe pulmonary disease in humans, dogs, rats, mice and other vertebrate species with acquired, induced, or inherited immune deficiency syndromes. In addition, this procedure will demonstrate Actinomyces and related species, Nocardia asteroids, and certain encapsulated bacteria.