Learn More
Scytek Laboratories Modified Gomori Methenamine-Silver Nitrate Stain (GMS Stain) Kit, Large
SDP
Catalog No. NC1969850
Encompass_Preferred
The Modified Gomori Methenamine-Silver Nitrate Stain (GMS Stain Kit) is intended for use in thehistologic visualization of fungi, basement membrane and some opportunistic organisms such asPneumocystis carinii. Pneumocystis carinii is an opportunistic pathogen that causes severe pulmonarydisease in humans, dogs, rats, mice and other vertebrate species with acquired, induced, or inheritedimmune deficiency syndromes. Fungi: BlackP. Carinii: BlackMucin: GrayMycelia (inner): Grey to BlackHyphae (inner): Grey to BlackBackground: Light GreenKit contents: SNS125, Silver Nitrate Solution (0.2%), 125 ml. MET125, Methenamine Solution, 125 ml. GCB125,Gold Chloride Solution, (0.2%) 125 ml. BOR015, Borax Solution, 15 ml. 18-25C.SBC125, Sodium Bisulfite Solution, 125 ml. CHR125, Chromic Acid Solution 125 ml. STB125, Sodium Thiosulfate Solution (5%), 125 ml. LGA125, Light Green Solution, 125 ml.
This item is not returnable. View return policy
Catalog No. NC1969850 Supplier Scytek Laboratories Supplier No. KAA1
The Modified Gomori Methenamine-Silver Nitrate Stain (GMS Stain Kit) is intended for use in thehistologic visualization of fungi, basement membrane and some opportunistic organisms such asPneumocystis carinii. Pneumocystis carinii is an opportunistic pathogen that causes severe pulmonarydisease in humans, dogs, rats, mice and other vertebrate species with acquired, induced, or inheritedimmune deficiency syndromes. Fungi: BlackP. Carinii: BlackMucin: GrayMycelia (inner): Grey to BlackHyphae (inner): Grey to BlackBackground: Light GreenKit contents: SNS125, Silver Nitrate Solution (0.2%), 125 ml. MET125, Methenamine Solution, 125 ml. GCB125,Gold Chloride Solution, (0.2%) 125 ml. BOR015, Borax Solution, 15 ml. 18-25C.SBC125, Sodium Bisulfite Solution, 125 ml. CHR125, Chromic Acid Solution 125 ml. STB125, Sodium Thiosulfate Solution (5%), 125 ml. LGA125, Light Green Solution, 125 ml.